Learn More
STEMCELL Technologies Papaïne
Click to view available options
Quantité:
25 mg
100 mg
Conditionnement:
100mg
Description
- Papain is a cysteine protease consisting of a single polypeptide containing three disulfide bridges
- Native crystalline papain is unreactive until acted upon by mild reducing agents, e.g. , cysteine, sulfide, or sulfite, and therefore likely exists as a zymogen
- Papain has a wide specificity with a preference towards arginine, lysine, and phenylalanine
- This enzyme degrades protein substrates such as the intercellular matrices of cartilage more extensively than pancreatic proteases and is typically less damaging and more effective than other proteases for tissue dissociation applications (Huettner & Baugham; Lam) and has also been used for the digestion of neural tissue (Fasano et al.)
Spécification
Spécification
| Description | Dopaminergic Neurons; Mesenchymal Cells, PSC-Derived; Neurons; Pancreatic Cells |
| Quantité | 100 mg |
| Synonyme | Papainase; Papaya peptidase I |
Correction du contenu d'un produit
Le programme Encompass de Fisher Scientific propose des articles qui ne font pas partie de notre portefeuille de distribution. Ces produits n'ont généralement pas de photos ou de descriptions détaillées. Cependant, nous nous engageons à améliorer votre expérience d'achat. Veuillez utiliser le formulaire ci-dessous pour nous faire part de vos commentaires sur le contenu de ce produit.