Learn More
DiscoverX PathHunter™ CHO-K1 GPR142 β-Arrestin Orphan GPCR Cell Line
Description
The PathHunter™ GPR142 cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand based activation of GPR142 GPCR activity via detection of β-arrestin recruitment. This product uses EFC technology (www.discoverx.com/technologies-platforms/enzyme-fragment-complementation-technology). The included cell line overexpresses PK-tagged GPR142 and EA-tagged β-Arrestin 2. Activation of GPR142 stimulates the recruitment of β-Arrestin 2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. Orphan receptor with unknown ligand.
Spécification
Spécification
| Entrée du dosage | β-Arrestin recruitment |
| Lignée cellulaire | CHO-K1 |
| À utiliser avec (application) | Cell Lines |
| Quantité | 2 flacons |
| Entrée cible | GPR142 |
| Numéro d’adhésion | NM_181790 |