Learn More
DiscoverX PathHunter™ eXpress IL4R/IL2RG Dimerization Assay
Click to view available options
Quantité:
200 ddp (2 x 96 puits)
Description
The PathHunter™ IL4R/IL2RG Dimerization eXpress kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting receptor-receptor interaction. This kit assesses ligand (e.g. IL-4) induced dimerization of IL4R/IL2RG receptors. This product uses EFC technology. The cell line included overexpresses IL4R target protein fused to PK alongside IL2RG target protein fused to EA. Upon ligand-induced dimerization of IL4R/IL2RG , detectable EFC signal is produced. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates.
Spécification
Spécification
| Entrée du dosage | IL4R/IL2RG Dimerization |
| À utiliser avec (application) | Assay-Ready eXpress Kits |
| Agoniste primaire | IL-4 |
| Quantité | 200 ddp (2 x 96 puits) |
| Entrée cible | IL4R/IL2RG |
| Lignée cellulaire | U2OS |
| Cible | IL4R/IL2RG |