missing translation for 'onlineSavingsMsg'
Learn More

PF 1022A, MedChemExpress

Code produit. 16461357
Encompass_Preferred
Click to view available options
Quantité:
5 mg
10 mg
50 mg
100 mg
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Code produit. 16461357

Marque: Medchem Express HY12361

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!

Cet article nécessite une mise à jour du prix avant d'être commandé.

Demandez un prix pour cet article ici.

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

MedChemExpress PF 1022A is a cyclooctadepsipeptide with broadspectrum anthelmintic properties produced by fermentation of the fungus Mycelia sterilia. PF 1022A is a channel-forming ionophore. PF 1022A showes strong anthelmintic activities against Ascaridia galli in chickens. PF 1022A also can be used for angiostrongyliasis research.

PF 1022A is a cyclooctadepsipeptide with broadspectrum anthelmintic properties produced by fermentation of the fungus Mycelia sterilia. PF 1022A is a channel-forming ionophore. PF 1022A showes strong anthelmintic activities against Ascaridia galli in chickens. PF 1022A also can be used for angiostrongyliasis research.

Specifications

Nom chimique ou matériau PF 1022A
Forme physique Solid
CAS 133413-70-4
Couleur Blanc
Quantité 50 mg
Notes de qualité de la pureté Research
Formule moléculaire C52H76N4O12
À utiliser avec (application) COVID-19-immunoregulation
Informations sur la solubilité DMSO : ≥ 43 mg/mL (45.30 mM)
SMILES O=C(N([C@H](C(O[C@](C(N([C@H](C(O[C@H](CC1=CC=CC=C1)C2=O)=O)CC(C)C)C)=O)([H])C)=O)CC(C)C)C)[C@H](OC([C@@H](N(C([C@H](OC([C@](N2C)([H])CC(C)C)=O)C)=O)C)CC(C)C)=O)CC3=CC=CC=C3
Poids moléculaire (g/mol) 949.18
Poids de la formule 949.18
Pourcentage de pureté 99.12%
Qualité Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.