missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Phosphofructokinase/PFK Activity Assay Kit (Colorimetric)

Codice prodotto. 18666771 Sfoglia Tutto Bio Techne Prodotti
Cambia vista
Click to view available options
Quantité:
96 tests
Dimensione della confezione:
1 kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantité unitSize
18666771 96 tests 1 kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 18666771 Fornitore Novus Biologicals™ N. del fornitore NBP325908

Please to purchase this item. Need a web account? Register with us today!

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Assay Kit (Colorimetric)

TRUSTED_SUSTAINABILITY

Specifica

Contenu et stockage Storage of components varies. See protocol for specific instructions.
Quantité 96 tests
Espèces Non-species specific
Titolo del prodotto
Selezionare un problema

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.