Learn More
BUCHI Pure Essential Chromatography System
Code produit. p-200136251 Tous les produits Buchi Produits
Click to view available options
Structure:
Avec détecteur
Sans détecteur
Conditionnement:
1 pièce
Règlementaire : Cet article est soumis à éco-contribution.
Code produit. 18087401
Marque: BUCHI 11C90010
Cet article nécessite une mise à jour du prix avant d'être commandé.
Demandez un prix pour cet article ici.
Règlementaire : Cet article est soumis à éco-contribution.
The Pure Essential Chromatography system focuses on the essentials for any basic flash application. The design of the instrument, its modularity, and user-friendliness make it easy for you to take the first steps into automation and profit from increased speed and endless possibilities.
Perform straightforward separations
Many flash separations only require a basic setup. At the bare minimum, a pump for smooth solvent and sample flow, and optionally a UV detector and fraction collector:
  • The Pure Essential Chromatography system benefits from a basic design and setup
  • The small footprint of the instrument allows for placement even in crowded labs, and its robustness makes it highly reliable
  • Basic but powerful and safe thanks to a 50 bar (725 psi) pump, pressure sensors, and regulators
A modular and scalable solution that grows with your needs
  • Every beginning is small. BUCHI supports you in every step of your research journey and offers the highest flexibility for your purification step:
  • Expansion possibilities include the addition of a UV detector and fraction collector to the Pure Chromatography C-900
  • There are no limits regarding the size of the collected fractions
  • Additional options allow you to be flexible regarding sample loading (liquid and solid) and the cartridge holder (any size between 4 to 5000 g)
Easy to use, plug and play chromatography
The Pure Essential Chromatography systems make flash separations easier than ever before; This is due to several key features related to the hardware and software:
  • Intuitive navigation provided by novel and state-of-the-art software (methods, run data, and general settings)
  • All tubing and parts of the instrument are easily accessible and allow the user to follow and monitor the process flow
  • Automation simplifies the entire separation process. It supports safe and efficient operation and reduces any worries about unforeseeable events
Spécification
Structure Avec détecteur
Type Flash/Prep HPLC/UV-Vis/ELSD