Learn More
Cole-Parmer PVC Lab and Vacuum Tubing
Tous les produits Cole-Parmer Produits
Click to view available options
Diamètre (impériale) interne:
0,0625 po
0,0781 po
0,125 po
0,188 po
0,25 po
0,2 po
0,313 po
0,5 po
0,75 po
15/64 po
1 po
3/32 po
3/8 in.
5/32 po
Diamètre (impériale) externe:
0,234 po
0,25 po
0,2 po
0,313 po
0,344 po
0,375 po
0,438 po
0,469 po
0,5 po
0,625 po
0,75 po
0,878 po
1 in
1,125 po
1,25 po
1/8 po
3/16 po
5/32 po
9 à 16 po
Conditionnement:
15m
Description
Versatile applications including peristaltic pumps, analytical instruments, incubators, gas, and drain lines. Glass-smooth inner bore design for easy clean-ups and changeovers, enhancing workflow efficiency. Ideal for general-purpose, laboratory, and single-use applications. Free technical application support available to assist with tubing selection
Spécification
Spécification
| Diamètre (impériale) interne | 5/32 po |
| Matériau | PVC |
| Diamètre (impériale) externe | 0,234 po |
| Longueur totale (impériale) | 50 ft. |
| Couleur | Translucide |
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.