Learn More
Hellma™ Cellule à flux UV en quartz
Click to view available options
Capacité (métrique):
31 μl
Conditionnement:
1 pièce
Specifications
Specifications
| Type | Cellule de flux UV |
| Capacité (métrique) | 31 μl |
| Matériau | Quartz |
| Longueur du trajet optique | 0,5 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction