Learn More
R&D Systems™ TACS-XL In Situ Apoptosis Detection Kits
Click to view available options
Méthode de détection:
Basique
DAB
Étiquette bleue
Conditionnement:
30 tests
Description
Includes
Proteinase K Solution; Cytonin; TdT enzyme, B-dNTP mix; Strep-HRP; anti-BrdU; Strep-Diluent
Based on incorporation of bromodeoxyuridine (BrdU) at the 3 ft. OH ends of the DNA fragments that form during apoptosis
This detection system uses a biotin conjugated anti-BrdU antibody and streptavidin-horseradish peroxidase for visualiza
Spécification
Spécification
| Quantité | 30 Tests |
| Type de produit | TACS-XL In Situ Apoptosis Detection Kit |
| Contenu et stockage | The product requires storage at -20 to -70° C, 2 - 8° C, and room temperature. Consult the product insert for specific storage temperatures. Do not use past expiration date. |
| Méthode de détection | Étiquette bleue |