Learn More
R&D Systems™ TACS-XL In Situ Apoptosis Detection Kits
Code produit. p-4830560
Click to view available options
Méthode de détection:
Basique
DAB
Étiquette bleue
Conditionnement:
30 tests
Code produit. 18491330
Marque: R&D Systems™ 482830BK
Includes
Proteinase K Solution; Cytonin; TdT enzyme, B-dNTP mix; Strep-HRP; anti-BrdU; Strep-Diluent
A TUNEL assay designed for fixed cells, embedded tissue, and frozen tissue using light and electron microscopy
Based on incorporation of bromodeoxyuridine (BrdU) at the 3 ft. OH ends of the DNA fragments that form during apoptosis
This detection system uses a biotin conjugated anti-BrdU antibody and streptavidin-horseradish peroxidase for visualiza
TRUSTED_SUSTAINABILITY
Spécification
Quantité 30 Tests
Type de produit TACS-XL In Situ Apoptosis Detection Kit
Contenu et stockage The product requires storage at -20 to -70° C, 2 - 8° C, and room temperature. Consult the product insert for specific storage temperatures. Do not use past expiration date.
Méthode de détection Étiquette bleue