Learn More
Invitrogen™ Rat anti-Mouse IgG1 (Heavy chain) Secondary Antibody, FITC
Code produit 15276029
Modifier l'affichage
Click to view available options
Quantité:
500 μg
Conditionnement:
500µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité Conditionnement
15276029 500 μg 500µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Code produit 15276029 Fournisseur Invitrogen™ Code fournisseur MA516785
  • ONLINE EXCLUSIVE PRICE
    666.00€
    619.38€ / 500µg
    Économisez 46.62€
    7% de réduction
 Article restreint
Expédition estimée: 14-12-2026
Rat Monoclonal Secondary Antibody
For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL. Rat anti Mouse IgG1 Heavy Chain antibody, clone LO-MG1-2 recognizes murine IgG1, and does not bind other mouse immunoglobulin classes or subclasses.
Anti-Mouse secondary antibodies are affinity-purified antibodies with well-characterized specificity for mouse immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
TRUSTED_SUSTAINABILITY
Spécification
Antigène Mouse IgG1 (Heavy chain)
Applications Flow Cytometry, Immunocytochemistry
Classification Monoclonal
Clone LO-MG1-2
Concentration 1 mg/mL
Conjugué FITC
Formule PBS with 50% glycerol and <0.1% sodium azide
Espèces hôtes Rat
Immunogène Purified murine IgG1 from BALB/c mice.
Méthode de purification Affinity chromatography
Quantité 500 μg
État réglementaire RUO
Primaire ou secondaire Secondary
Espèces cibles Mouse
Contenu et stockage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark.
Type de produit Secondary Antibody
Forme Liquid
Isotype IgG1