missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Rat CXCL12 (SDF-1a) Recombinant Protein, PeproTech®
Click to view available options
Quantité:
1 mg
10 μg
100 μg
2 μg
250 μg
50 μg
500 μg
Conditionnement:
100µg
10µg
1mg
250µg
2µg
500µg
50µg
Description
Recombinant Rat SDF-1alpha is a 7.9 kDa protein containing 68 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis
CXCL12 is a stromal cell-derived alpha chemokine member of the intercrine family. This gene product and its receptor CXCR4 can activate lymphocytes and have been implicated in the metastasis of some cancers such as breast cancer. Mutations in this gene are associated with resistance to human immunodeficiency virus type 1 infections. Multiple transcript variants encoding different isoforms have been found for this gene.
Spécification
Spécification
| À utiliser avec (application) | ELISA, Functional Assay, Western Blot |
| Formule | protein with no preservative |
| Identification génétique (Entrez) | 24772 |
| Poids moléculaire | 7.9 kDa |
| Nom | Rat CXCL12 (SDF-1 A) |
| Quantité | 1 mg |
| Source | E. coli |
| État réglementaire | RUO |
| Concentration en endotoxines | <1 EU/ μg |
| Alias de gène | 12-O-tetradecanoylphorbol 13-acetate repressed protein 1; C Cmotif chemokine; C x C motif chemokine; CC motif chemokine; CCmotif chemokine; chemokine (C-X-C motif) ligand 12; chemokine (C-X-C motif) ligand 12 (stromal cell-derived factor 1); CXC; CXC motif chemokine; C-X-C motif chemokine 12; C-X-C motif chemokine ligand 12; CXCL; Cxcl12; H-SDF-1-alpha; Intercrine reduced in hepatomas; IRH; Pbsf; pre-B cell growth-stimulating factor; pre-B-cell growth-stimulating factor; SCYB12; SDF 1 A; SDF 1 alpha; SDF 1 A; SDF1; SDF-1; SDF1 alpha; SDF-1 gamma; SDF1A; SDF-1-alpha(3-67); SDF1B; SDF-1-beta(3-72); SDF1a; Stromal cell-derived factor 1; stromal cell-derived factor-1 alpha; stromal cell-derived factor-1 gamma; thymic lymphoma cell-stimulating factor; TLSF; Tpar1 |
| Afficher plus |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu