missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Rat IL-13 (113 aa) Recombinant Protein, PeproTech®
Click to view available options
Quantité:
1 mg
10 μg
100 μg
2 μg
250 μg
500 μg
Conditionnement:
100µg
10µg
1mg
250µg
2µg
500µg
Description
Recombinant Rat IL-13 a 12.3 kDa protein consisting of 113 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis
IL-13 is an immunoregulatory cytokine produced primarily by activated Th2 cells, and is involved in regulating inflammatory and immune responses. IL-13 down-regulates macrophage activity, thereby inhibiting the production of pro-inflammatory cytokines and chemokines, and is involved in allergen-induced asthma through mechanisms independent of IgE and eosinophils. IL-13 also regulates several stages of B cell maturation and differentiation, up-regulates CD23 and MHC class II expression, and promotes IgE isotype switching of B cells. Structurally, IL-13 closely resembles IL-3, IL-4, IL-5 and GM-CSF. The gene encoding IL-13, IL3, IL5, IL4, and CSF2 form a cytokine gene cluster on chromosome 5q, with the IL-13 gene particularly close to IL4.
Spécification
Spécification
| Numéro d’adhésion | P42203 |
| À utiliser avec (application) | ELISA, Functional Assay, Western Blot |
| Formule | protein with no preservative |
| Identification génétique (Entrez) | 116553 |
| Poids moléculaire | 12.3 kDa |
| Nom | Rat IL-13 (113 aa) |
| Quantité | 1 mg |
| Source | E. coli |
| État réglementaire | RUO |
| Concentration en endotoxines | <1 EU/ μg |
| Afficher plus |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu