missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Rat IL-2, Animal-Free Recombinant Protein, PeproTech®
Click to view available options
Quantité:
1 mg
100 μg
20 μg
500 μg
Conditionnement:
100µg
1mg
20µg
500µg
Description
Recombinant Rat IL-2 is a 15.3 kDa protein containing 134 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis
Interleukin 2 (IL-2) is an immuno-modulatory cytokine that is important for the proliferation of activated T cells, differentiation of B cells, natural killer cells, monocytes and macrophages. IL-2 signals through the IL-2 receptor (IL-2R), a heterotrimeric protein complex whose gamma chain is also shared by interleukin 4 (IL-4) and interleukin 7 (IL-7). The expression of the IL-2 gene in mature thymocytes is monoallelic, which represents an unusual regulatory mode for controlling the precise expression of a single gene. The targeted disruption of a gene similar to IL-2 in mice leads to an ulcerative colitis-like disease that suggests an essential role of this gene in the immune response to antigenic stimuli.
Spécification
Spécification
| Numéro d’adhésion | P17108 |
| À utiliser avec (application) | Functional Assay |
| Formule | protein with no preservative |
| Identification génétique (Entrez) | 116562 |
| Poids moléculaire | 15.3 kDa |
| Nom | Rat IL-2, Animal-Free |
| Quantité | 500 μg |
| Source | E. coli |
| État réglementaire | RUO |
| Concentration en endotoxines | <0.1 EU/ μg |
| Afficher plus |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu