Learn More
R&D Systems™ Rat Luminex™ Discovery Assay
Click to view available options
Quantité:
11 Plex
1 Plex
2 Plex
4 Plex
8 Plex
Conditionnement:
1 kit
1 pièce
Spécification
Spécification
| Quantité | 1 Plex |
| Espèces | Rat |
| Cible de liaison | Multiplex Cytokine Assay Kits |
| Contenu et stockage | Store the unopened kit at 2°C to 8°C. Do not use past the expiration date on the label. Avoid freezing microparticles. Protect microparticles from light. |