missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Canine LRG1 Protein

Code produit. 15980889
Click to view available options
Quantité:
10 μg
100 μg
2 μg
Conditionnement:
100µg
10µg
2µg
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Code produit. 15980889

Marque: enQuireBio™ QP125822ug

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!

This item is not returnable. View return policy

A cDNA sequence encoding the LRG1 was constructed and used to recombinantly synthesize the protein.

Specifications

Identification génétique (Entrez) 611876
Nom LRG1 Protein
Quantité 2 μg
État réglementaire Research Use Only
Concentration en endotoxines Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Symbole de gène(s) LRG1
Type de produit Recombinant Protein
Réactivité croisée Canine
Espèces Canine
Marqueur de protéine Untagged
Séquence MSPWSPERSQ SPGGLSSHLS KTLLLLLFSV ASAQGVTPDR DTCLVFPSSN GSSVSCHPPA KLPHHFPSDT VHLVVEFFNL TQLRPDTLQG VSNLQELHLS TNQLESLSPK LLLPVPLLKV LDLTRNALTQ LPPGLFRVSA ALHTLVLKEN RLEVLEPSWL CGLKALGHLD LSGNHLQTLP PGLLANVTAL RILDLSNNQL KTLPLDLLRG PLQLERLHLE GNRLQVLEEG LLAPQPGLRY LFLSDNKLAA VAAGAFRGLQ QLDMLDLSNN SLTSVPKGLW TSLGQPTRDM EDGFDISGNP WICDHNLDDL YRWLVANKDK MFSRNDTRCA RPEALKGQTL LEAAESH
Tampon LRG1 protein filtered (0.4microm) and lyophilized in 0.5 mg/ml in 20mM TRIS and 50mM NaCl, pH 8.0.
Pureté ou qualité Purity greater than 90% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.