missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant E. Coli IDNK Protein

A cDNA sequence encoding the IDNK was constructed and used to recombinantly synthesize the protein.

Marque:  enQuireBio™ QP12368-1mg

Informations supplémentaires : Poids : 0.01000kg

 Afficher plus de versions de ce produit

Code produit. 15980189

  • 3740.00€ / 1mg
Expédition estimée: 11-07-2023
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Spécification

Spécification

IDNK Protein
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
E. coli
His
IDNK protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 10% glycerol and 1mM DTT.
Greater than 95% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMAGESFI LMGVSGSGKT LIGSKVAALL SAKFIDGDDL HPAKNIDKMS QGIPLSDEDR LPWLERLNDA SYSLYKKNET GFIVCSSLKK QYRDILRKGS PHVHFLWLDG DYETILARMQ RRAGHFMPVA LLKSQFEALE RPQADEQDIV RIDINHDIAN VTEQCRQAVL AIRQNRICAK EGSASDQRCE
Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions