missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. Coli MUG Protein
Click to view available options
Quantité:
1 mg
10 μg
2 μg
Conditionnement:
10µg
1mg
2µg
Spécification
Spécification
| Identification génétique (Entrez) | 947560 |
| Nom | MUG Protein |
| Quantité | 1 mg |
| État réglementaire | Research Use Only |
| Concentration en endotoxines | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
| Symbole de gène(s) | mug |
| Type de produit | Recombinant Protein |
| Réactivité croisée | E. coli |
| Espèces | E. coli |
| Marqueur de protéine | His |
| Afficher plus |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction