missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant E. Coli NDK Protein

A cDNA sequence encoding the NDK was constructed and used to recombinantly synthesize the protein.

Marque:  enQuireBio™ QP12807-20ug

Informations supplémentaires : Poids : 0.01000kg

 Afficher plus de versions de ce produit

Code produit. 15981599

  • 189.00€ / 20µg
Expédition estimée: 18-07-2023
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Spécification

Spécification

NDK Protein
20 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
E. coli
His
NDK protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 10% glycerol, 0.1M NaCl and 1mM DTT.
Greater than 95% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSHMAIERT FSIIKPNAVA KNVIGNIFAR FEAAGFKIVG TKMLHLTVEQ ARGFYAEHDG KPFFDGLVEF MTSGPIVVSV LEGENAVQRH RDLLGATNPA NALAGTLRAD YADSLTENGT HGSDSVESAA REIAYFFGEG EVCPRTR
Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions