missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Feline IL-5 Protein
Click to view available options
Quantité:
10 μg
Conditionnement:
10µg
Description
The Recombinant Feline IL-5 Protein is derived from NS0. The Recombinant Feline IL-5 Protein has been validated for the following applications: Bioactivity.
Spécification
Spécification
| Formule | Lyophilized from a 0.2μm filtered solution in PBS. |
| Identification génétique (Entrez) | 493803 |
| Poids moléculaire | M.W.Observed: 12-14 kDa and 18-22 kDa, under reducing conditions |
| Nom | IL-5 |
| Quantité | 10 μg |
| Source | Mouse myeloma cell line,NS0-derived feline IL-5 protein Ile20-Ser134,with a C-terminal 10-His tag |
| Conditions de stockage | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Forme structurelle | Disulfide-linked homodimer |
| Conjugué | Unconjugated |
| Recombinant | Recombinant |
| Afficher plus |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu