Learn More
enQuireBio™ Recombinant Hepatitis C HCV NS3 Genotype-6a Protein
A cDNA sequence encoding the HCV NS3 Genotype-6a was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP12174-100ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
Hepatitis C HCV NS3 Genotype-6a Protein | |
100 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Hepatitis C | |
His |
HCV NS3 Genotype-6a protein is >95% pure as determined by 10% PAGE (coomassie staining). | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
1.5M urea, 25mM Tris-HCl pH 8.0, 0.2% Triton-X and 50% Glycerol. |