Learn More
enQuireBio™ Recombinant Human BLyS / TNFSF13B / BAFF Protein
Recombinant Protein
Marque:  enQuireBio™ QP5331-1mg
Informations supplémentaires : Poids : 0.01000kg
Code produit. 15985919
  • 7105.00€ / 1mg
Expédition estimée: 17-01-2024
Spécification
NP_001139117.1
BLyS / TNFSF13B / BAFF Protein
Research Use Only
TNFSF13B
Human
Untagged
10 mM sodium phosphate, pH 7.5
10673
1 mg
Less than or equal to 1.00 EUs/ug as determined by the LAL method.
Recombinant Protein
E. coli
MHHHHHHLVP RAVQGPEETV TQDCLQLIAD SETPTIQKGS YTFVPWLLSF KRGSALEEKE NKILVKETGY FFIYGQVLYT DKTYAMGHLI QRKKVHVFGD ELSLVTLFRC IQNMPETLPN NSCYSAGIAK LEEGDELQLA IPRENAQISL DGDVTFFGAL KLL
Greater than 90 as determined by SDS-PAGE.