Learn More
enQuireBio™ Recombinant Human CD1B Protein
A cDNA sequence encoding the CD1B was constructed and used to recombinantly synthesize the protein.
131.00€ - 1740.00€
Spécification
Identification génétique (Entrez) | 910 |
---|---|
Nom | CD1B Protein |
État réglementaire | Research Use Only |
Concentration en endotoxines | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Symbole de gène(s) | CD1B |
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
15926798
|
enQuireBio™
QP11309-2UG |
2 μg |
131.00€
2µg |
Estimated Shipment: 27-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15916798
|
enQuireBio™
QP11309-10UG |
10 μg |
203.00€
10µg |
Estimated Shipment: 27-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15906798
|
enQuireBio™
QP11309-100UG |
100 μg |
1740.00€
100µg |
Estimated Shipment: 27-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Spécification
910 | |
Research Use Only | |
CD1B | |
Human | |
His | |
CD1B protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.2M NaCl, 20% glycerol and 1mM DTT. |
CD1B Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMSEHAFQ GPTSFHVIQT SSFTNSTWAQ TQGSGWLDDL QIHGWDSDSG TAIFLKPWSK GNFSDKEVAE LEEIFRVYIF GFAREVQDFA GDFQMKYPFE IQGIAGCELH SGGAIVSFLR GALGGLDFLS VKNASCVPSP EGGSRAQKFC ALIIQYQGIM ETVRILLYET CPRYLLGVLN AGKADLQRQV KPEAWLSSGP SPGPGRLQLV CHVSGFYPKP VWVMWMRGEQ EQQGTQLGDI LPNANWTWYL RATLDVADGE AAGLSCRVKH SSLEGQDIIL YWRNPTSIGS | |
Greater than 90.0% as determined by SDS-PAGE. |