missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human CD44 (CHO-expressed) Fc Protein, CF
Click to view available options
:
50 μg
Description
Measured by its binding ability in a functional ELISA. Recombinant Human CD44 Fc Chimera (Catalog # 11622-CD) binds to Recombinant Human LSECtin/CLEC4G Protein (Catalog # 2947-CL ) with a ED50 of 5.00-50.0 ng/mL.
Spécification
Spécification
| À utiliser avec (application) | Bioactivity |
| Formule | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. |
| Identification génétique (Entrez) | 960 |
| Poids moléculaire | M.W.-theoretical: 96 kDa, M.W.-observed: > 190 kDa, under non-reducing conditions. |
| Quantité | 50 μg |
| Source | Chinese Hamster Ovary cell line, CHO-derived human CD44 protein Human CD44 (Gln21-Trp650) (N-terminus) Accession # P16070.3 GGIEGRMD Human IgG1 (Pro100-Lys330) (C-terminus) |
| Conditions de stockage | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution. |
| Concentration en endotoxines | <0.10 EU per 1 μg of the protein by the LAL method. |
| Alias de gène | CD44, CD44 antigen, CD44 molecule (Indian blood group), CD44R, CDw44, cell surface glycoprotein CD44, chondroitin sulfate proteoglycan 8, CSPG8, ECMR-III, Epican, Extracellular matrix receptor III, GP90 lymphocyte homing/adhesion receptor, HCAM, HCELL, hematopoietic cell E- and L-selectin ligand, Heparan sulfate proteoglycan, Hermes antigen, homing function and Indian blood group system, HUTCH-I, Hyaluronate receptor, IN, LHR, MC56, MDU2, MDU2CD44 antigen (homing function and Indian blood group system), MDU3, MDU3CDW44, MIC4, MIC4MGC10468, MUTCH-I, Pgp1, PGP-1, PGP-I, Phagocytic glycoprotein 1, Phagocytic glycoprotein I |
| Conjugué | Unconjugated |
| Afficher plus |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu