missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human CD74 Protein, CF
Click to view available options
:
50 μg
Description
Measured by its binding ability in a functional ELISA. Recombinant Human CD74 (Catalog # 3590-CDB) binds to Human CD74 Antibody (Catalog # AF3590 ) with an ED50 of 0.300-3.60 ng/mL.
Spécification
Spécification
| Numéro d’adhésion | NP_004346.1 |
| À utiliser avec (application) | Bioactivity |
| Formule | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. |
| Identification génétique (Entrez) | 972 |
| Poids moléculaire | M.W.-theoretical: 19 kDa, M.W.-observed: 25-33 kDa, under reducing conditions. |
| Quantité | 50 μg |
| Source | Chinese Hamster Ovary cell line, CHO-derived human CD74 protein Gln73-Met232, with an N-terminal HA (YPYDVPDYA) tag |
| Conditions de stockage | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution. |
| Concentration en endotoxines | <0.10 EU per 1 μg of the protein by the LAL method. |
| Alias de gène | CD74, CD74 antigen, CD74 antigen (invariant polypeptide of major histocompatibility complex, classII antigen-associated), CD74 molecule, major histocompatibility complex, class II invariant chain, CLIP, DHLAG, DHLAGgamma chain of class II antigens, HLA class II histocompatibility antigen gamma chain, HLADG, HLA-DR antigens-associated invariant chain, HLA-DR-gamma, Ia antigen-associated invariant chain, Ia-associated invariant chain, Ia-GAMMA, Ii, INVG34, MHC HLA-DR gamma chain, p33 |
| Afficher plus |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu