missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Human Centaurin alpha 1 GST (N-Term) Protein

Product Code. 18603611 Shop All Bio Techne Products
Change view
Click to view available options
Quantité:
10 μg
25 μg
Unit Size:
10µg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantité unitSize
18603611 25 μg 25µg
18655081 10 μg 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18603611 Supplier Novus Biologicals™ Supplier No. H00011033P0125ug

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!

This item is not returnable. View return policy

Recombinant Protein

This protein was produced in an in vitro wheat germ expression system that should preserve correct conformational folding that is necessary for biological function. While it is possible that this protein could display some level of activity, the functionality of this protein has not been explicitly measured or validated.
TRUSTED_SUSTAINABILITY

Specifications

À utiliser avec (application) Western Blot, ELISA, Protein Array, Immunoaffinity Purification
Formule 50 mM Tris-HCl, 10 mM reduced Glutathione, pH 8.0 in the elution buffer.
Identification génétique (Entrez) 11033
Poids moléculaire MolecularWeight-theroretical: 69.8 kDa
Quantité 25 μg
Conditions de stockage Store at -80°C. Avoid freeze-thaw cycles.
Alias de gène arf-GAP with dual PH domain-containing protein 1, ArfGAP with dual PH domains 1, CENTA1centaurin-alpha-1, centaurin, alpha 1, centaurin-alpha, Centaurin-alpha-1, Cnt-a1, GCS1L, p42IP4, Putative MAPK-activating protein PM25
Symbole de gène(s) ADAP1
Catégorie de recherche Signal Transduction
Pureté ou qualité >80% by SDS-PAGE and Coomassie blue staining
Protéine Centaurin alpha 1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.