missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human CILP-1 Protein
Click to view available options
:
50ug; Carrier Free
Description
The Recombinant Human CILP-1 Protein is derived from NS0. The Recombinant Human CILP-1 Protein has been validated for the following applications: Bioactivity.
Spécification
Spécification
| Formule | Lyophilized from a 0.2μm filtered solution in PBS. |
| Identification génétique (Entrez) | 8483 |
| Poids moléculaire | 78.8 kDa |
| Nom | CILP-1 |
| Quantité | 50μg |
| Source | Mouse myeloma cell line,NS0-derived human CILP-1 protein Met1-Arg720,with a C-terminal 6-His tag |
| Conditions de stockage | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Conjugué | Unconjugated |
| Recombinant | Recombinant |
| Pureté ou qualité | 95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu