missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Clathrin interactor 1 Protein
A DNA sequence encoding the Homo sapiens (Human) Clathrin interactor 1, was expressed in the hosts and tags indicated.
Marque: enQuireBio™ QP5839-ec-200ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
NP_001182484.1 | |
Clathrin interactor 1 Protein | |
Research Use Only | |
CLINT1 | |
Recombinant Protein | |
E. coli | |
MLNMWKVREL VDKATNVVMN YSEIESKVRE ATNDDPWGPS GQLMGEIAKA TFMYEQFPEL MNMLWSRMLK DNKKNWRRVY KSLLLLAYLI RNGSERVVTS AREHIYDLRS LENYHFVDEH GKDQGINIRQ KVKELVEFAQ DDDRLREERK KAKKNKDKYV GVSSDSVGGF RYSERYDPEP KSKWDEEWDK NKSAFPFSDK LGELSDKIGS TIDDTISKFR RKDREDSPER CSDSDEEKKA RRGRSPKGEF KDEEETVTTK HIHITQATET TTTRHKRTAN PSKTIDLGAA AHYTGDKASP DQNASTHTPQ SSVKTSVPSS KSSGDLVDLF DGTSQSTGGS ADLFGGFADF GSAAASGSFP SQVTATSGNG DFGDWSAFNQ APSGPVASSG EFFGSASQPA VELVSGSQSA LGPPPAASNS SDLFDLMGSS QATMTSSQSM NFSMMSTNTV GLGLPMSRSQ NTDMVQKSVS KTLPSTWSDP SVNISLDNLL PGMQPSKPQQ PSLNTMIQQQ NMQQPMNVMT QSFGAVNLSS PSNMLPVRPQ TNALIGGPMP MSMPNVMTGT MGMAPLGNTP MMNQSMMGMN MNIGMSAAGM GLTGTMGMGM PNIAMTSGTV QPKQDAFANF ANFSK | |
Greater than 90% as determined by SDS-PAGE. |
9685 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |