missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CST3 Protein

Product Code. 15994248
Click to view available options
Quantité:
1 mg
10 μg
2 μg
Unit Size:
10µg
1mg
2µg
This item is not returnable. View return policy

Product Code. 15994248

Brand: enQuireBio™ QP105612ug

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!

This item is not returnable. View return policy

A cDNA sequence encoding the CST3 was constructed and used to recombinantly synthesize the protein.

Specifications

Nom CST3 Protein
Quantité 2 μg
État réglementaire Research Use Only
Concentration en endotoxines < 1.0 EU per ug protein as determined by the LAL method.
Activité biologique The inhibitory function of Cystatin-C on papin’s protease activity was measured by a colorimetric assay using L-BAPA as substrate. IC50 value was measured at 5-20 microg/ml (0.3-1.5 microm) with a range of 1.56-50microg/ml Cystatin-C in presence of 0.55microm papain and 0.44microm L-BAPA. The activity is typically 0.3-1.5microm IC50.
Type de produit Recombinant Protein
Réactivité croisée Human
Espèces Human (HEK293)
Marqueur de protéine Untagged
Tampon The Cystatin-C was lyophilized from 1 mg/ml in 1xPBS.
Pureté ou qualité Greater than 95% as obsereved by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.