Learn More
enQuireBio™ Recombinant Human DNAJC12 Protein
Code produit. 15998008
Click to view available options
Quantité:
1 mg
10 μg
2 μg
Conditionnement:
10µg
1mg
2µg
Code produit. 15998008
Marque: enQuireBio™ QP116782ug
A cDNA sequence encoding the DNAJC12 was constructed and used to recombinantly synthesize the protein.
Specifications
Nom DNAJC12 Protein
Quantité 2 μg
État réglementaire Research Use Only
Concentration en endotoxines Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Type de produit Recombinant Protein
Réactivité croisée Human
Espèces E. coli
Marqueur de protéine His
Séquence MGSSHHHHHH SSGLVPRGSH MGSHMDAILN YRSEDTEDYY TLLGCDELSS VEQILAEFKV RALECHPDKH PENPKAVETF QKLQKAKEIL TNEESRARYD HWRRSQMSMP FQQWEALNDS VKTSMHWVVR GKKDLMLEES DKTHTTKMEN EECNEQRERK KEELASTAEK TEQKEPKPLE KSVSPQNSDS SGFADVNGWH LRFRWSKDAP SELLRKFRNY EI
Tampon The DNAJC12 solution (0.5 mg/ml) contains 20mM Tris-HCl buffer, pH 8.0, 10% glycerol, 2mM DTT and 100mM NaCl.
Pureté ou qualité Greater than 90% as determined by SDS-PAGE.