Learn More
enQuireBio™ Recombinant Human EEF1G Protein
A cDNA sequence encoding the EEF1G was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP11749-20ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
EEF1G Protein | |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The EEF1G solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 10% glycerol and 1mM DTT. |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMAAGTLY TYPENWRAFK ALIAAQYSGA QVRVLSAPPH FHFGQTNRTP EFLRKFPAGK VPAFEGDDGF CVFESNAIAY YVSNEELRGS TPEAAAQVVQ WVSFADSDIV PPASTWVFPT LGIMHHNKQA TENAKEEVRR ILGLLDAYLK TRTFLVGERV TLADITVVCT LLWLYKQVLE PSFRQAFPNT NRWFLTCINQ PQFRAVLGEV KLCEKMAQFD AKKFAETQPK KDTPRKEKGS REEKQKPQAE RKEEKKAAAP APEEEMDECE QALAAEPKAK DPFAHLPKST FVLDEFKRKY SNEDTLSVAL PYFWEHFDKD GWSLWYSEYR FPEELTQTFM SCNLITGMFQ RLDKLRKNAF ASVILFGTNN SSSISGVWVF RGQELAFPLS PDWQVDYESY TWRKLDPGSE ETQTLVREYF SWEGAFQHVG KAFNQGKIFK |