Learn More
enQuireBio™ Recombinant Human Epiplakin Protein
A DNA sequence encoding the Homo sapiens (Human) Epiplakin, was expressed in the hosts and tags indicated.
Marque: enQuireBio™ QP5991-ec-100ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
NP_112598.3 | |
Epiplakin Protein | |
100 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
83481 | |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
EPPK1 | |
Recombinant Protein | |
E. coli | |
MSGHTLPPLP VPGTNSTEQA SVPRAMAATL GAGTPPRPQA RSIAGVYVEA SGQAQSVYAA MEQGLLPAGL GQALLEAQAA TGGLVDLARG QLLPVSKALQ QGLVGLELKE KLLAAERATT GYPDPYGGEK LALFQAIGKE VVDRALGQSW LEVQLATGGL VDPAQGVLVA PEPACHQGLL DRETWHKLSE LEPGTGDLRF LNPNTLERLT YHQLLERCVR APGSG |