missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human Erythropoietin/EPO Protein
Click to view available options
Quantité:
1 mg
10 μg
250 μg
50 μg
Conditionnement:
10µg
1mg
250µg
50µg
Description
Measured in a cell proliferation assay using TF-1 human erythroleukemic cells. Kitamura, T. et al. (1989) J. Cell Physiol. 140:323. The ED50 for this effect is 0.075-0.750 ng/mL.
Spécification
Spécification
| Numéro d’adhésion | CAA26094.1 |
| À utiliser avec (application) | Bioactivity |
| Formule | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. |
| Poids moléculaire | M.W.-observed: 33-41 kDa, under reducing conditions., M.W.-theroretical: 21 kDa |
| Quantité | 250 μg |
| Source | Chinese Hamster Ovary cell line, CHO-derived human Erythropoietin/EPO protein Ala28-Arg193 |
| Conditions de stockage | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. |
| Concentration en endotoxines | <0.10 EU / 1 μg of the protein by the LAL method. |
| Conjugué | Unconjugated |
| Reconstitution | Reconstitute at 100-500 μg/mL in PBS. |
| Afficher plus |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu