missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human GFR alpha-1/GDNF R alpha-1 His Protein
Click to view available options
Quantité:
100 μg
Conditionnement:
100µg
Description
Measured by its binding ability in a functional ELISA. Recombinant Human GFR alpha-1/GDNF R alpha-1 His-tag (Catalog # 11398-GR) binds Recombinant Human GDNF Protein (Catalog # 212-GD/CF ) with an ED50 of 26.0-260 ng/mL.
Spécification
Spécification
| Numéro d’adhésion | P56159.2 |
| À utiliser avec (application) | Bioactivity |
| Formule | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. |
| Identification génétique (Entrez) | 2674 |
| Poids moléculaire | MolecularWeight-observed: 51-64 kDa, under reducing conditions., MolecularWeight-theroretical: 46 kDa |
| Quantité | 100 μg |
| Source | Human embryonic kidney cell, HEK293-derived human GFR alpha-1/GDNF R alpha-1 protein Ala19-Ser429, with a C-terminal 6-His tag |
| Conditions de stockage | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. |
| Concentration en endotoxines | <0.10 EU / 1 μg of the protein by the LAL method. |
| Alias de gène | DKFZp313E1012, DKFZp686J0156, GDNF family receptor alpha 1, GDNF family receptor alpha-1, GDNF R alpha-1, GDNF receptor alpha-1, GDNFRAGFR-ALPHA-1, GDNFR-alpha-1, GDNFRMGC23045, GFR alpha1, GFRa-1, GFRA1, GFR-alpha-1, Glial cell line-derived neurotrophic factor receptor alpha, GPI-linked anchor protein, PI-linked cell-surface accessory protein, RET ligand 1, RET1L, RETL1FLJ10561, TGF-beta related neurotrophic factor receptor 1, TGF-beta-related neurotrophic factor receptor 1, TRNR1FLJ31546 |
| Afficher plus |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu