missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Human HORMAD2 GST (N-Term) Protein
Tous les produits Bio Techne ProduitsDescription
This protein was produced in an in vitro wheat germ expression system that should preserve correct conformational folding that is necessary for biological function. While it is possible that this protein could display some level of activity, the functionality of this protein has not been explicitly measured or validated.
Specifications
Specifications
| À utiliser avec (application) | Western Blot, ELISA, Protein Array, Immunoaffinity Purification |
| Formule | 50 mM Tris-HCl, 10 mM reduced Glutathione, pH 8.0 in the elution buffer. |
| Identification génétique (Entrez) | 150280 |
| Poids moléculaire | MolecularWeight-theroretical: 61.7 kDa |
| Quantité | 10 μg |
| Conditions de stockage | Store at -80°C. Avoid freeze/thaw cycles. |
| Alias de gène | HORMA domain containing 2, HORMA domain-containing protein 2, MGC26710 |
| Symbole de gène(s) | HORMAD2 |
| Pureté ou qualité | >80% by SDS-PAGE and Coomassie blue staining |
| Protéine | HORMAD2 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?