missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human HSP70/HSPA1A Protein
Click to view available options
Quantité:
100 μg
50 μg
Conditionnement:
100µg
50µg
Description
Reaction conditions will need to be optimized for each specific application. We recommend an initial HSP70/HSPA1A concentration of 2-3 μM for in vitro use. IMPORTANT: HSP40/DNAJB1 (Catalog # AP-110), or another suitable co-chaperone, is required for HSP70/HSPA1A activity and should be used at a concentration that is equimolar to HSP70/HSPA1A.
Spécification
Spécification
| Formule | X mg/mL (X μM) in 50 mM HEPES pH 8, 100 mM NaCl, 5 mM DTT |
| Identification génétique (Entrez) | 3303 |
| Poids moléculaire | 70 kDa |
| Nom | HSP70/HSPA1A |
| Quantité | 100 μg |
| Source | E. coli-derived human HSP70/HSPA1A protein |
| Conditions de stockage | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -70° C as supplied. 3 months, -70° C under sterile conditions after opening. |
| Forme structurelle | Solution |
| Conjugué | Unconjugated |
| Recombinant | Recombinant |
| Afficher plus |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu