missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human IGF-II R/IGF2R (aa 43-1365) Protein
Click to view available options
Quantité:
50 μg
Conditionnement:
50µg
Description
The Recombinant Human IGF-II R/IGF2R (aa 43-1365) Protein is derived from CHO. The Recombinant Human IGF-II R/IGF2R (aa 43-1365) Protein has been validated for the following applications: Bioactivity.
Spécification
Spécification
| Numéro d’adhésion | P11717.3 |
| Formule | Lyophilized from a 0.2μm filtered solution in PBS. |
| Identification génétique (Entrez) | 3482 |
| Poids moléculaire | 147 kDa (monomer) |
| Nom | IGF-II R |
| Quantité | 50 μg |
| Source | Chinese Hamster Ovary cell line, CHO-derived human IGF-II R/IGF2R protein Ala43-Asp1365, with a C-terminal 6-His tag |
| Conditions de stockage | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Forme structurelle | Noncovalently-linked homodimer |
| Conjugué | Unconjugated |
| Afficher plus |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu