missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IL33 / Interleukin-33 / NF-HEV Protein
Click to view available options
Quantité:
10 μg
100 μg
2 μg
250 μg
Unit Size:
100µg
10µg
250µg
2µg
Specifications
Specifications
| Numéro d’adhésion | NP_001186569.1 |
| Identification génétique (Entrez) | 90865 |
| Nom | IL33 / Interleukin-33 / NF-HEV Protein |
| Quantité | 2 μg |
| État réglementaire | Research Use Only |
| Concentration en endotoxines | Less than 1.0 EU/ug as determined by LAL method. |
| Symbole de gène(s) | IL33 |
| Longueur des protéines | Partial (see sequence information for more details). |
| Activité biologique | The ED50 as determined by a cell proliferation assay using murine D10S cells is less than 0.05 ng/ml, corresponding to a specific activity of > 2.0 M 107 IU/mg. |
| Type de produit | Recombinant Protein |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction