Learn More
enQuireBio™ Recombinant Human KCNMB3 Protein
Code produit. p-7150343
Click to view available options
Quantité:
1 mg
20 μg
5 μg
Conditionnement:
1mg
20µg
5µg
Code produit. 15990549
Marque: enQuireBio™ QP124795ug
A cDNA sequence encoding the KCNMB3 was constructed and used to recombinantly synthesize the protein.
Spécification
Nom KCNMB3 Protein
Quantité 5 μg
État réglementaire Research Use Only
Concentration en endotoxines Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Type de produit Recombinant Protein
Réactivité croisée Human
Espèces E. coli
Marqueur de protéine His
Séquence MGSSHHHHHH SSGLVPRGSH MGSKPFMLSI QREESTCTAI HTDIMDDWLD CAFTCGVHCH GQGKYPCLQV FVNLSHPGQK ALLHYNEEAV QINPKCFYTP KCHQDRNDLL NSALDIKEFF DHKNGTPFSC FYSPASQSED VILIKKYDQ
Tampon KCNMB3 protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.4M UREA and 10% glycerol.
Pureté ou qualité Greater than 85.0% as determined by SDS-PAGE.