missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human L1CAM Fc Chimera Protein
Click to view available options
:
100ug; Carrier Free
Description
The Recombinant Human L1CAM Fc Chimera Protein is derived from NS0. The Recombinant Human L1CAM Fc Chimera Protein has been validated for the following applications: Bioactivity.
Spécification
Spécification
| Formule | Lyophilized from a 0.2μm filtered solution in Tris-Citrate and NaCl. |
| Identification génétique (Entrez) | 3897 |
| Poids moléculaire | 150.4 kDa (monomer) & 55.9 kDa (truncated) |
| Nom | L1CAM Fc Chimera |
| Quantité | 100μg |
| Source | Mouse myeloma cell line,NS0-derived human L1CAM protein, (N-terminus) Human L1CAM (Ile20-Glu1120) & (Arg864-Glu1120) Accession &Num; CAA42508 IEGRMD Human IgG1 (Pro100-Lys330) 6-His tag (C-terminus) |
| Conditions de stockage | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 4 weeks, -20 to -70° C under sterile conditions after reconstitution. |
| Forme structurelle | Disulfide-linked homodimer |
| Conjugué | Unconjugated |
| Recombinant | Recombinant |
| Afficher plus |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu