missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human LFA 3 Protein

A cDNA sequence encoding the LFA 3 was constructed and used to recombinantly synthesize the protein.

Marque:  enQuireBio™ QP12556-1mg

Informations supplémentaires : Poids : 0.01000kg

 Afficher plus de versions de ce produit

Code produit. 15980799

  • 1185.00€ / 1mg
Expédition estimée: 10-07-2023
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Spécification

Spécification

965
Greater than 98.0% as determined by(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE.
Research Use Only
CD58
Human
Untagged
Each mg of CD58 contains 0.8 mg sucrose, 0.3 mg glycine, 0.25 mg sodium citrate dihydrate, and 4microg citric acid monohydrate.
LFA 3 Protein
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
CHO
MVAGSDAGRA LGVLSVVCLL HCFGFISCFS QQIYGVVYGN VTFHVPSNVP LKEVLWKKQK DKVAELENSE FRAFSSFKNR VYLDTVSGSL TIYNLTSSDE DEYEMESPNI TDTMKFFLYV LESLPSPTLT CALTNGSIEV QCMIPEHYNS HRGLIMYSWD CPMEQCKRNS TSIYFKMEND LPQKIQCTLS NPLFNTTSSI ILTTCIPSSG HSRHRYALIP IPLAVITTCI VLYMNGILKC DRKPDRTNSN
Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions