Learn More
enQuireBio™ Recombinant Human LRG1 Protein
Code produit. p-7150455
Click to view available options
Quantité:
1 mg
20 μg
5 μg
Conditionnement:
1mg
20µg
5µg
Code produit. 15940899
Marque: enQuireBio™ QP12583HIS5ug
A cDNA sequence encoding the LRG1 was constructed and used to recombinantly synthesize the protein.
Spécification
Nom LRG1 Protein
Quantité 5 μg
État réglementaire Research Use Only
Concentration en endotoxines Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Type de produit Recombinant Protein
Réactivité croisée Human
Espèces E. coli
Marqueur de protéine His
Séquence VTLSPKDCQV FRSDHGSSIS CQPPAEIPGY LPADTVHLAV EFFNLTHLPA NLLQGASKLQ ELHLSSNGLE SLSPEFLRPV PQLRVLDLTR NALTGLPPGL FQASATLDTL VLKENQLEVL EVSWLHGLKA LGHLDLSGNR LRKLPPGLLA NFTLLRTLDL GENQLETLPP DLLRGPLQLE RLHLEGNKLQ VLGKDLLLPQ PDLRYLFLNG NKLARVAAGA FQGLRQLDML DLSNNSLASVPEGLWASLGQ PNWDMRDGFD ISGNPWICDQ NLSDLYRWLQ AQKDKMFSQN DTRCAGPEAV KGQTLLAVAK SQ
Tampon LRG1 protein filtered (0.4microm) and lyophilized from 20mM Tris and 20mM NaCl, pH 8.
Pureté ou qualité Greater than 95.0% as determined by SDS-PAGE.