missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Midkine / MDK Protein
Click to view available options
Quantité:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Conditionnement:
100µg
10µg
1mg
200µg
500µg
50µg
Specifications
Specifications
| Numéro d’adhésion | NP_001012333.1 |
| Identification génétique (Entrez) | 4192 |
| Nom | Midkine / MDK Protein |
| Quantité | 200 μg |
| État réglementaire | Research Use Only |
| Concentration en endotoxines | Not determined. |
| Symbole de gène(s) | MDK |
| Longueur des protéines | Full Length |
| Type de produit | Recombinant Protein |
| Réactivité croisée | Human |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction