Learn More
enQuireBio™ Recombinant Human MLLT11 Protein
Code produit. 15901279
Click to view available options
Quantité:
1 mg
20 μg
5 μg
Conditionnement:
1mg
20µg
5µg
Code produit. 15901279
Marque: enQuireBio™ QP127015ug
A cDNA sequence encoding the MLLT11 was constructed and used to recombinantly synthesize the protein.
Spécification
Nom MLLT11 Protein
Quantité 5 μg
État réglementaire Research Use Only
Concentration en endotoxines Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Type de produit Recombinant Protein
Réactivité croisée Human
Espèces E. coli
Marqueur de protéine His
Séquence MGSSHHHHHH SSGLVPRGSH MGSMRDPVSS QYSSFLFWRM PIPELDLSEL EGLGLSDTAT YKVKDSSVGK MIGQATAADQ EKNPEGDGLL EYSTFNFWRA PIASIHSFEL DLL
Tampon MLLT11 protein solution (0.25 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.4M Urea and 10% glycerol.
Pureté ou qualité Greater than 80.0% as determined by SDS-PAGE.