missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human MMADHC Protein
Click to view available options
Quantité:
1 mg
20 μg
5 μg
Conditionnement:
1mg
20µg
5µg
Specifications
Specifications
| Nom | MMADHC Protein |
| Quantité | 20 μg |
| État réglementaire | Research Use Only |
| Concentration en endotoxines | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
| Type de produit | Recombinant Protein |
| Réactivité croisée | Human |
| Espèces | E. coli |
| Marqueur de protéine | His |
| Séquence | MGSSHHHHHH SSGLVPRGSH MGSSDESHVA AAPPDICSRT VWPDETMGPF GPQDQRFQLP GNIGFDCHLN GTASQKKSLV HKTLPDVLAE PLSSERHEFV MAQYVNEFQG NDAPVEQEIN SAETYFESAR VECAIQTCPE LLRKDFESLF PEVANGKLMI LTVTQKTKND MTVWSEEVEI EREVLLEKFI NGAKEICYAL RAEGYWADFI DPSSGLAFFG PYTNNTLFET DERYRHLGFS VDDLGCCKVI RHSLWGTHVV VGSIFTNATP DSHIMKKLSG N |
| Tampon | The MMADHC solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 1mM DTT and 10% glycerol. |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction