Learn More
enQuireBio™ Recombinant Human NMB Protein
A cDNA sequence encoding the NMB was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP12853-1mg
Informations supplémentaires : Poids : 0.01000kg
Spécification
NMB Protein | |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The NMB solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.4M urea and 10% glycerol. |
Greater than 85.0% as determined by SDS-PAGE. | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSAPLSWDL PEPRSRASKI RVHSRGNLWA TGHFMGKKSL EPSSPSPLGT APHTSLRDQR LQLSHDLLGI LLLKKALGVS LSRPAPQIQY RRLLVQILQK |