Learn More
enQuireBio™ Recombinant Human OSTF1 Protein
A cDNA sequence encoding the OSTF1 was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP12936-50ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
OSTF1 Protein | |
50 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
OSTF1 protein (1 mg/ml) solution contains 20mM Tris-HCl buffer pH 8.0, 0.5mM DTT and 10% glycerol. |
Greater than 95.0% as determined by SDS-PAGE. | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MSKPPPKPVK PGEGGQVKVF RALYTFEPRT PDELYFEEGD IIYITDMSDT NWWKGTSKGR TGLIPSNYVA EQAESIDNPL HEAAKRGNLS WLRECLDNRV GVNGLDKAGS TALYWACHGG HKDIVEMLFT QPNIELNQQN KLGDTALHAA AWKGYADIVQ LFLAKGARTD LRNIEKKLAF DMATNAACAS LLKKKQGTDA VRTLSNAEDY LDDEDSDLEH HHHHH |