missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human PDK1 Protein

A cDNA sequence encoding the PDK1 was constructed and used to recombinantly synthesize the protein.

Marque:  enQuireBio™ QP12998-1mg

Informations supplémentaires : Poids : 0.01000kg

 Afficher plus de versions de ce produit

Code produit. 15982189

  • 2490.00€ / 1mg
Expédition estimée: 10-07-2023
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Spécification

Spécification

PDK1 Protein
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
PDK1 solution containing 20mM Tris pH 7, 0.5mM DTT, 0.1M NaCl, 0.1mM EDTA 0.1mM PMSF, 1mM MgCl2 and 40% glycerol.
Greater than 95.0% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MSSDSGSSPA SERGVPGQVD FYARFSPSPL SMKQFLDFGS VNACEKTSFM FLRQELPVRL ANIMKEISLLPDNLLRTPSV QLVQSWYIQS LQELLDFKDK SAEDAKAIYD FTDTVIRIRN RHNDVIPTMA QGVIEYKESF GVDPVTSQNV QYFLDRFYMS RISIRMLLNQ HSLLFGGKGK GSPSHRKHIG SINPNCNVLE VIKDGYENAR RLCDLYYINS PELELEELNA KSPGQPIQVV YVPSHLYHMVFELFKNAMRA TMEHHANRGV YPPIQVHVTL GNEDLTVKMS DRGGGVPLRK IDRLFNYMYS TAPRPRVETS RAVPLAGFGY GLPISRLYAQ YFQGDLKLYS LEGYGTDAVI YIKALSTDSI ERLPVYNKAA WKHYNTNHEA DDWCVPSREP KDMTTFRSA
Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions