Learn More
enQuireBio™ Recombinant Human PIH1D2 Protein
A cDNA sequence encoding the PIH1D2 was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP13053-10ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
120379 | |
Greater than 85.0% as determined by SDS-PAGE. | |
Research Use Only | |
PIH1D2 | |
Human | |
His | |
PIH1D2 protein solution (0.25 mg/ml) containing Phosphate buffered saline (pH 7.4), 20% glycerol and 1mM DTT. |
PIH1D2 Protein | |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMETSSKG LLTQVTQFWN LLDDLAQSDP EGYEKFIQQQ LKEGKQLCAA PEPQLCLQTR ILKPKEKILF INLCQWTRIP APQSTTHPVP LTVGKPEDTT EISDAYTVID VAYNPDVLHA AEKDQVKKNQ LIQMAMKCIE EKFQFTLSHS YHITKFRIKG SIQRMKQNLM GIQTDSIDLR EKMRRELTLG QIRSSTMSNP DHFPQLLLPK DQVSGKAVCL IEEISSTEIQ VEMKMPAYEL KIVHDHSEKP LKIELKVELP GINSVSLCDL SVSEDDLLIE VSEKYRLHLN LPKLIDTEMT TAKFIKEKST LIITMPLV |