Learn More
enQuireBio™ Recombinant Human RPS2 Protein
Code produit. p-7151240
Click to view available options
Quantité:
1 mg
20 μg
5 μg
Conditionnement:
1mg
20µg
5µg
Code produit. 15903309
Marque: enQuireBio™ QP133545ug
A cDNA sequence encoding the RPS2 was constructed and used to recombinantly synthesize the protein.
Spécification
Nom RPS2 Protein
Quantité 5 μg
État réglementaire Research Use Only
Concentration en endotoxines Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Type de produit Recombinant Protein
Réactivité croisée Human
Espèces E. coli
Marqueur de protéine His
Séquence MGSSHHHHHH SSGLVPRGSH MGSMADDAGA AGGPGGPGGP GMGNRGGFRG GFGSGIRGRG RGRGRGRGRG RGARGGKAED KEWMPVTKLG RLVKDMKIKS LEEIYLFSLP IKESEIIDFF LGASLKDEVL KIMPVQKQTR AGQRTRFKAF VAIGDYNGHV GLGVKCSKEV ATAIRGAIIL AKLSIVPVRR GYWGNKIGKP HTVPCKVTGR CGSVLVRLIP APRGTGIVSA PVPKKLLMMA GIDDCYTSAR GCTATLGNFA KATFDAISKT YSYLTPDLWK ETVFTKSPYQ EFTDHLVKTH TRVSVQRTQA PAVATT
Tampon RPS2 protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0),10% glycerol and 0.4M Urea.
Pureté ou qualité Greater than 85% as determined by SDS-PAGE.